missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
Marca: Brady™ N003263
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifica
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
Pipe Markers - Eau chaude sanitaire | |
White on Green | |
Polyester | |
Eau chaude sanitaire | |
5 | |
5 Markers/Card | |
-40°F to +194°F | |
0.0049 in. | |
5.91 in. | |
0.47 in. |
EAUCH.SANIT.150X12CARD-T1-P16 | |
Permanent Rubber, Permanent, Pressure-sensitive | |
Card | |
Pipe Marker | |
Pipe and valve marking | |
French | |
5 Pk. | |
-40°C to +90°C | |
0.124 mm | |
150 mm | |
12 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto