Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
Marca:  Brady™ N002811
 vedi altre versioni di questo prodotto
Codice prodotto. 12579737
  • 13.50€ / pezzi 5
In stock
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifica
Specifica
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Pipe Markers - Fuel
White on Dark Brown
Card
Pipe Marker
Pipe and valve marking
French
5 Pk.
-40°C to +90°C
0.124 mm
150 mm
12 mm
FUEL150X12CARD-T1-P19
Permanent Rubber, Permanent, Pressure-sensitive
Warning
Polyester
Fuel
5
5 Markers/Card
-40°F to +194°F
0.0049 in.
5.91 in.
0.47 in.