Learn More
Eppendorf™ Pipette Holder 2
Eppendorf™ Pipette Holder 2 keeps your Eppendorf™ pipette within good reach and saves precious bench space.
Marca: Eppendorf™ 3116000112
33.92 EUR valido fino al 2025-04-30
Usa il codice promo "24138" per avere il tuo prezzo promozionale.
Descrizione
Eppendorf™ Pipette Holder 2 keeps your Eppendorf™ pipette within good reach and saves precious bench space.
- Carries one manual single or multi-channel pipette including Eppendorf Research™, Research™ plus, Reference™, Reference™ 2 or Biomaster™
- Designed for wall mounting or for installation in Pipette Carousel 2 and Charger Carousel 2
- Sticky tape for wall mounting included
Specifica
Sticky tape for wall mounting | |
For wall mounting or for installation in Pipette Carousel 2 and Charger Carousel 2 | |
One manual single or multi-channel pipette including Eppendorf Research™, Research™ plus, Reference™, Reference™2 or Biomaster™ |
Offerte speciali
-
Diversi produttori
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsCodice sconto: 25721Scade: 2025-05-01
nuova offerta Save Now
Titolo del prodotto
Eppendorf™ Pipette Holder 2 > For use with: One manual single or multi-channel p