Learn More
Brady™ Etichette in poliestere stampabili con trasferimento termico
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Marca: Brady™ 217079
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Glossy white finish with permanent acrylic is thermal transfer printable
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -94 to 230° F (-70 to 110° C)
Specifica
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted | |
Thermal Transfer Printable Labels | |
White | |
Polyester | |
Thermal Transfer Printer Label | |
BBP11, BP1244, BP1344, BP4000, BP4320 | |
3 | |
Rectangle | |
-75°C to +145°C | |
21.98 mm | |
80.58 mm |
BPT-616-488 | |
Acrylic, Permanent | |
Matte | |
Thermal Transfer | |
Laboratory identification | |
25.86 mm | |
3000/Pk. | |
22.86W x 19 mmH | |
0.022 mm | |
22.86 mm | |
19.05 mm |