Learn More
Masterflex™ Polyethylene Barbed Elbow Connector
Create ninety-degree bends in flexible tubing without the kinks
Marca: Masterflex™ 30622-67
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifica
Barbed Elbow Connector | |
Not Autoclavable | |
2 1/32 in. | |
3/4 in. | |
3/4 in. | |
2 1/32 in. |
Barbed Connection | |
HDPE | |
3/4 in. | |
3/4 in. | |
Ethylene Oxide | |
Translucent |