Learn More
Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles
Click to view available options
Capacity: 500mL; Closure Color: Blue
Descrizione
Wide mouth design aids easy and safe filling of contents.
- Made from polyethylene
- Color-coded caps for easy identification
- Ingenious cap designed to avoid involuntary dripping
- Suitable for storing food stuff
Specifica
Specifica
Capacità (sistema metrico) | 500 mL |
Include | Color coded cap |
Materiale | Polyethylene |
Dimensioni filettatura (sistema metrico) | GL45 |
Colore chiusura | Blue |
Graduazioni | 100mL |
Graduated | Sì |
Bocca | Wide |
Titolo del prodotto
Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles >