Learn More
Applied Biosystems™ POP-7™ Polymer, for 3500/SeqStudio™ Flex
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
Marca: Applied Biosystems™ 4393708
263.64 EUR valido fino al 2025-05-31
Usa il codice promo "24127" per avere il tuo prezzo promozionale.
Includes: The pouch contains 384 samples of the polymer. Store the pouch at 2 to 8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches cannot be used on the instrument.
Descrizione
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
• Easy to use: pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part⁄lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: allow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID⁄Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
Each
Specifica
384 Samples | |
Long-Read Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing) | |
POP-7™ |
Certificati
Per poter ottenere i certificati è necessario indicare un numero di lotto. Per individuare il numero di lotto su ordini precedenti usi l'area dedicata allo stato degli ordini.
Numero di lotto | Tipo di certificato | Data | Catalog Number |
---|---|---|---|
2411322 | Certificato di analisi | 17/01/2025 | 4393708 |
2410321 | Certificato di analisi | 31/12/2024 | 4393708 |
2410320 | Certificato di analisi | 23/12/2024 | 4393708 |
2410319 | Certificato di analisi | 12/12/2024 | 4393708 |
2410318 | Certificato di analisi | 11/11/2024 | 4393708 |
Offerte speciali
-
Diversi produttori
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Codice sconto: 24127Scade: 2025-05-31
nuova offerta Save Up to 70%