Learn More
Rogo-Sampaic Imbuto Buchner in porcellana
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
Marca: Rogo-Sampaic PNF009
Descrizione
- White porcelain
- Available for a range of volumes and filter diameters
- Applications include collecting recrystalized compounds
- Excellent thermal and chemical resistance
Specifica
80 ml | |
55 mm | |
64 mm | |
Imbuto Buchner |
Porcellana | |
55 mm | |
1 ciascuno |