Learn More
Gilson™ Position Rack Tube Vessel Type, Fraction Collector
These racks are for use with the FC203B and FC204 Fraction Collectors
Marca: Gilson™ 130412
Dettagli aggiuntivi : Peso : 0.45000kg
Descrizione
- HPLC Purification Technique
- Maximum Vessel Quantity is 80
- Tube Vessel Type
Specifica
Code 4 | |
5 mL Vial (12 x 75 mm) | |
80 |