missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone Receptor Ab-8 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.
Marca: Epredia MS-298-RQ
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.Host Species: Mouse
Clone: hPRa 2 + hPRa 3
Isotype: IgG1/κ + IgG1/κ
Species Reactivity: Human, Horse, Sheep and Pig. Others not known.
Epitope: N-terminal half of human PgR
Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (B-form) and 81kDa (A-form)
Positive Control: T47D or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Specifica
Progesterone Receptor Ab-8 | |
Monoclonal | |
Unconjugated | |
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice | |
ASR, IVD | |
Primary |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
hPRa 2+hPRa 3 | |
Mouse | |
12 mL | |
Cancer and Tumor Biology | |
Ready-to-Use |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto