Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Marca: Quickfit GC10
30.21 EUR valido fino al 2025-01-31
Usa il codice promo "23894" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Highly resistant to chemical attack
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifica
Joint Clip | |
Encapsulated Steel spring | |
10/19 |
PTFE | |
Conical Joints | |
250°C |