Learn More
Cowie Technology™ PTFE Straight Through Connector
Accepts any rigid or semi-rigid tubing. Cowie Technology™ PTFE Straight Through Connector locks in place quickly with a bayonet type coupling.
Marca: Cowie Technology™ 016.902.5.2
Dettagli aggiuntivi : Peso : 0.00700kg
Descrizione
- Bayonet fitting
- For use with rigid and semi rigid tubing such as glass, FEP or PTFE
Specifica
PTFE Straight Through Connector | |
Autoclavable | |
PTFE | |
-200°C | |
14 mm | |
White |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
4.5 mm | |
44 mm | |
1 Each | |
14 mm | |
280°C |