missing translation for 'onlineSavingsMsg'
Learn More
Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals Codice prodotto.: 11400643

Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals

Codice prodotto. 11400643
2500 g
Click to view available options
Quantità:
100 g
500 g
2500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11400643

Marca: Thermo Scientific Alfa Aesar A12712.0E

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.

Solubility
Soluble in dimethyl sulfoxide, acetone, chloroform, ethyl ether, n-hexane and benzene.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 89-32-7
Formula molecolare C10H2O6
Molecular Weight (g/mol) 218.12
Numero MDL MFCD00005005
InChI Key ANSXAPJVJOKRDJ-UHFFFAOYSA-N
Sinonimo pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydride
PubChem CID 6966
IUPAC Name furo[3,4-f][2]benzofuran-1,3,5,7-tetrone
SMILES O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O

Specifica

Materiale o nome chimico Pyromellitic dianhydride
Punto di fusione 284°C to 288°C
Densità 1.68
Punti di ebollizione 305°C to 310°C (30 mmHg)
Punto d'infiammabilità 380°C (716°F)
Quantità 2500 g
Beilstein 213583
Sensibilità Moisture sensitive
Informazioni di solubilità Soluble in dimethyl sulfoxide,acetone,chloroform,ethyl ether,n-hexane and benzene.
Peso formulazione 218.12
Percent Purity 97%
Vedi altri risultati Mostra meno risultati

Certificati

Per poter ottenere i certificati è necessario indicare un numero di lotto. Per individuare il numero di lotto su ordini precedenti usi l'area dedicata allo stato degli ordini.

5 Risultati trovati
Numero di lotto Tipo di certificato Data Catalog Number
Numero di lotto10252686 Tipo di certificatoCertificato di analisi Data07/10/2024 Catalog NumberA12712.0E,A12712.22,A12712.36
Numero di lotto10250312 Tipo di certificatoCertificato di analisi Data10/04/2024 Catalog NumberA12712.0E,A12712.22,A12712.36
Numero di lotto10213461 Tipo di certificatoCertificato di analisi Data20/06/2022 Catalog NumberA12712.0E,A12712.22,A12712.36
Numero di lotto10239665 Tipo di certificatoCertificato di analisi Data12/04/2022 Catalog NumberA12712.0E,A12712.22,A12712.36
Numero di lottoUSLY000195 Tipo di certificatoCertificato di analisi Data22/11/2021 Catalog NumberA12712.0E,A12712.22,A12712.36
compliance-icons compliance-icons
Identificativo prodotto
  • Pyromellitic dianhydride
Avvertenza
  • Pericolo
Categoria di pericolo
  • Lesione oculare grave/irritazione oculare Categoria 1
  • Sensibilizzante delle vie respiratorie Categoria 1
  • Sensibilizzazione della pelle Categoria 1
Indicazioni di pericolo
  • H317-Può provocare una reazione allergica cutanea.
  • H318-Provoca gravi lesioni oculari.
  • H334-Può provocare sintomi allergici o asmatici o difficoltà respiratorie se inalato.
Consigli di prudenza
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P310-Contattare immediatamente un CENTRO ANTIVELENI/un medico .
  • P342+P311-In caso di sintomi respiratori: contattare un CENTRO ANTIVELENI/un medico/ .
Informazioni supplementari
  • MIXTURE LIST-Contiene: Pyromellitic dianhydride

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato