Learn More
Macherey Nagel Qualitative Analysis Test Papers, MN 615
Widely used for routine separations.
Marca: Macherey Nagel 4310015
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Qualitative filter papers widely used for routine separations
- Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
- Most widely used general purpose filter paper with medium retention and flow rate
- This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifica
Disc | |
MN 615 | |
100/Pk. | |
15 mm |
Titolo del prodotto
Macherey Nagel Qualitative Analysis Test Papers, MN 615 > Diameter: 15 mm