Learn More
Sartorius Qualitative Grade 1291 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Marca:  Sartorius FT-3-209-125
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10052431
  • 20.70€ / pezzi 100
Spedizione stimata: 04-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifica
Specifica
Disc
180 sec.
100/Pk.
125 mm
2 to 3 μm
1291
0.16 mm