missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat heart tissue lysate, Non-denatured; Abnova
Marca: Abnova L036W2.200ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Western Blotting, Immunoprecipitation
Specifica
Immunoprecipitation, Western Blot | |
Whole tissue lysate (non-denatured) | |
Tissue lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was adjusted to 5 mg/ml. | |
200 μg | |
Rat | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Heart | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In modified RIPA Lysis Buffer. | |
5 mg/mL |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Rat heart tissue lysate, Non-denatured; Abnova > 50μg
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto