missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat spleen tissue lysate, Denatured; Abnova
Marca: Abnova L042W1.200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Western Blotting
Especificaciones
Western Blot | |
Whole tissue lysate (denatured) | |
Tissue lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was adjusted to 3.75 mg/ml, and then mixed with 5X Sample Buffer to become final 3 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. | |
200 μg | |
Rat | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Spleen | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). | |
3 mg/mL |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Rat spleen tissue lysate, Denatured; Abnova > 50μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido