missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAPDH Protein
A DNA sequence encoding the Homo sapiens (Human) GAPDH, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8304-ec-1mg
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_001243728.1 | |
GAPDH Protein | |
Research Use Only | |
GAPDH | |
Recombinant Protein | |
E. coli | |
KVKVGVNGFG RIGRLVTRAA FNSGKVDIVA INDPFIDLNY MVYMFQYDST HGKFHGTVKA ENGKLVINGN PITIFQERDP SKIKWGDAGA EYVVESTGVF TTMEKAGAHL QGGAKRVIIS APSADAPMFV MGVNHEKYDN SLKIISNASC TTNCLAPLAK VIHDNFGIVE GLMTTVHAIT ATQKTVDGPS GKLWRDGRGA LQNIIPASTG AAKAVGKVIP ELNGKLTGMA FRVPTANVSV VDLTCRLEKP AKYDDIKKVV KQASEGPLKG ILGYTEHQVV SSDFNSDTHS STFDAGAGIA LNDHFVKLIS WYDNEFGYSN RVVDLMAHMA SKE | |
Greater than 90% as determined by SDS-PAGE. |
2597 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |