missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB8 Protein
A cDNA sequence encoding the RAB8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13247-1mg
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
RAB8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAKTYDY LFKLLLIGDS GVGKTCLLFR FSEDAFNTTF ISTIGIDFKI RTIELDGKKI KLQIWDTAGQ ERFRTITTAY YRGAMGIMLV YDITNEKSFD NIKNWIRNIE EHASSDVERM ILGNKCDMND KRQVSKERGE KLAIDYGIKF LETSAKSSAN VEEAFFTLAR DIMTKLNRKM NDSNSAGAGG PVKITENRSK KTSFFRC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |