missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPA1 Protein
A DNA sequence encoding the Homo sapiens (Human) RPA1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6618-ec-100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_002936.1 | |
RPA1 Protein | |
Research Use Only | |
RPA1 | |
Recombinant Protein | |
E. coli | |
VGQLSEGAIA AIMQKGDTNI KPILQVINIR PITTGNSPPR YRLLMSDGLN TLSSFMLATQ LNPLVEEEQL SSNCVCQIHR FIVNTLKDGR RVVILMELEV LKSAEAVGVK IGNPVPYNEG LGQPQVAPPA PAASPAASSR PQPQNGSSGM GSTVSKAYGA SKTFGKAAGP SLSHTSGGTQ SKVVPIASLT PYQSKWTICA RVTNKSQIRT WSNSRGEGKL FSLELVDESG EIRATAFNEQ VDKFFPLIEV NKVYYFSKGT LKIANKQFTA VKNDYEMTFN NETSVMPCED DHHLPTVQFD FTGIDDLENK SKDSLVDIIG ICKSYEDATK ITVRSNNREV AKRNIYLMDT SGKVVTATLW GEDADKFDGS RQPVLAIKGA RVSDFGGRSL SVLSSSTIIA NPDIPEAYKL RGWFDAEGQA LDGVSISDLK SGGVGGSNTN WKTLYEVKSE NLGQGDKPDY FSSVATVVYL RKENCMYQAC PTQDCNKKVI DQQNGLYRCE KCDTEFPNFK YRMILSVNIA DFQENQWVTC FQESAEAILG QNAAYLGELK DKNEQAFEEV FQNANFRSFI FRVRVKVETY NDESRIKATV MDVKPVDYRE YGRRLVMSIR RSALM | |
Greater than 90% as determined by SDS-PAGE. |
6117 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |