missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TRP2 Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Marca: Novus Biologicals™ NBP2-53051-250UG
1336.00 EUR valido fino al 2025-03-28
Usa il codice promo "25306" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 24-472 of Human TRP2 The Recombinant Human TRP2 Protein is derived from Baculovirus. The Recombinant Human TRP2 Protein has been validated for the following applications: SDS-Page.
Specifica
SDS-PAGE | |
1638 | |
Human TRP2 Protein | |
250 μg | |
QFPRVCMTVD SLVNKECCPR LGAESANVCG SQQGRGQCTE VRADTRPWSG PYILRNQDDR ELWPRKFFHR TCKCTGNFAG YNCGDCKFGW TGPNCERKKP PVIRQNIHSL SPQEREQFLG ALDLAKKRVH PDYVITTQHW LGLLGPNGTQ PQFANCSVYD FFVWLHYYSV RDTLLGPGRP YRAIDFSHQG PAFVTWHRYH LLCLERDLQR LIGNESFALP YWNFATGRNE CDVCTDQLFG AARPDDPTLI SRNSRFSSWE TVCDSLDDYN HLVTLCNGTY EGLLRRNQMG RNSMKLPTLK DIRDCLSLQK FDNPPFFQNS TFSFRNALEG FDKADGTLDS QVMSLHNLVH SFLNGTNALP HSAANDPIFV VLHSFTDAIF DEWMKRFNPP ADAWPQELAP IGHNRMYNMV PFFPPVTNEE LFLTSDQLGY SYAIDLPVSV EETPGWPTTH HHHHH | |
RUO | |
dopachrome delta-isomerase, dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2), DT, EC 5.3.3.12, L-dopachrome tautomerase, TRP2, TRP-2, Tyrosinase-related protein 2, TYRP2L-dopachrome Delta-isomerase | |
Recombinant Protein | |
Human |
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol | |
52.1kDa | |
>90%, by SDS-PAGE | |
Baculovirus | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
<1.0 EU per 1μg of protein (determined by LAL method) | |
DCT | |
Unconjugated | |
Recombinant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Novus Biologicals™ Recombinant Human TRP2 Protein > Quantity: 250μg
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto