Learn More
Thermo Scientific™ Remel™ Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated)
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
Marca: Thermo Scientific™ R111884
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
- Settling Plates (Single Bag and Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
-
Specifica
Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 | |
Monoplate - Double + Bag | |
Agar | |
10/Pk. |
Not intended for IVD use.