Learn More
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Marca:  Mettler Toledo™ 51108751
Codice prodotto. 12655815
  • 3475.00€ / Pezzo
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Coulometric Cells
  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Coulometric Cell with Diaphragm
Ketones, aldehydes, very low water values or where best possible accuracy is needed
Glass