Learn More
Brady™ Inserti marcafili Rigid DuraSleeve per M710
Inserti marcafili Rigid DuraSleeve per M710
Marca: Brady™ 312004
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Brady™ DuraSleeve™ printable inserts create a durable legend
- Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
- Legend can be changed before or after termination
Specifica
M7-23x4.4-7697-WT | |
White | |
Label | |
Wire and Cable Markers | |
Data and Telecommunications Labeling, Electrical Labeling, Wire and Cable Labeling | |
1 Label | |
Rectangle | |
-40°C to +120°C | |
0.35 mm | |
0.173 in. | |
23 mm |
Non-adhesive | |
Opaco | |
Polyester | |
Non-sterile | |
M710, BMP71 | |
500 Labels | |
-40°F to +248°F | |
0.0138 in. | |
4.4 x 23 mm | |
0.906 in. |