Learn More
Abnova™ RRP22 Recombinant Protein
Marca: Abnova™ H00010633-P01.10ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
AAH58077 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
48.07 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 μg | |
MGGSLRVAVLGAPGVGKTAIIRQFLFGDYPERHRPTDGPRLYRPAVLLDGAVYDLSIRDGDVAGPGSSPGGPEEWPDAKDWSLQDTDAFVLVYDICSPDSFDYVKALRQRIAETRPAGAPEAPILVVGNKRDRQRLRFGPRRALAALVRRGWRCGYLECSAKYNWHVLRLFRELLRCALVRARPAHPALRLQGALHPARCSLM | |
RRP22 | |
RASL10A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10633 | |
RRP22 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RASL10A | |
Human | |
Recombinant | |
Solution |