missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RRP22 Recombinant Protein

344.00€ - 521.00€

Specifications

Numero di accesso AAH58077
Da utilizzare con (applicazione) Antibody Production, Protein Array, ELISA, Western Blot
Formulazione 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
ID gene (immissione) 10633
Peso molecolare 48.07
View More Specs

Products 2
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
16115462
missing translation for 'viewDocuments'
Abnova™
H00010633-P01.10ug
10 μg
344.00€
10µg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16125462
missing translation for 'viewDocuments'
Abnova™
H00010633-P01.25ug
25 μg
521.00€
25µg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH58077
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
48.07
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RASL10A
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
10633
RRP22 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MGGSLRVAVLGAPGVGKTAIIRQFLFGDYPERHRPTDGPRLYRPAVLLDGAVYDLSIRDGDVAGPGSSPGGPEEWPDAKDWSLQDTDAFVLVYDICSPDSFDYVKALRQRIAETRPAGAPEAPILVVGNKRDRQRLRFGPRRALAALVRRGWRCGYLECSAKYNWHVLRLFRELLRCALVRARPAHPALRLQGALHPARCSLM
RRP22
RASL10A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.