Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
GREENER_CHOICE
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Marca:  Thermo Scientific™ 191NL
37.90 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Codice prodotto. 15397803
  • 38.30€ / pezzi 500
In stock
per visualizzare lo stock disponibile
Edge
Esplora altre offerte speciali
Aggiungi al carrello
Descrizione
Descrizione
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
  • Shatterproof, nontoxic polyethylene
  • Nonsterile
  • Low affinity surface
  • Freezer ready
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Polyethylene
250 μL
3 mm
Non-sterile
10.19 cm
Videos
  • Thermo Scientific Samco - Exact volume tranfer pipettes
    Video Player is loading.
    Current Time 0:00
    Duration 1:33
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:33
    1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    Thermo Scientific Samco Exact volume tranfer pipettes
  • Thermo Scientific Samco - Exact volume tranfer pipettes
    Video Player is loading.
    Current Time 0:00
    Duration 1:33
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:33
    1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    Thermo Scientific Samco Exact volume tranfer pipettes
SDS
Documenti Certificati
Titolo del prodotto
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes > 250 μL