Learn More
Thermo Scientific™ Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
3607.95€
Specifica
Descrizione | Refrigerated Vapor Cold Trap RVT450 |
---|---|
Tipo | Refrigerated Vapor Trap |
Codice del prodotto | Marca | Descrizione | Requisiti elettrici | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Descrizione | Requisiti elettrici | Prezzo | Quantità e disponibilità | |||||
15996161
|
Thermo Scientific™
RVT450-230 |
Refrigerated Vapor Cold Trap RVT450 | 230 V 50 Hz |
3607.95€
Pezzo |
Spedizione stimata: 13-05-2025
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT). Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump.
- Rapid cooling time minimizes waiting period
- Easy to remove glass flask for quick solvent changes
- Rapidly cools to operating temperature
- Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
- Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
Specifica
Refrigerated Vapor Cold Trap RVT450 | |
Refrigerated Vapor Trap |