Learn More
SB 204990, MedChemExpress
MedChemExpress SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.
Marca: Medchem Express HY-16450
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.Specifica
SB 204990 | |
White | |
C18H22Cl2O5 | |
DMSO : ≥ 100 mg/mL (256.89 mM) ∣H2O : 25 mg/mL (64.22 mM; Need ultrasonic) | |
389.27 | |
95.35% | |
Solid |
154566-12-8 | |
Research | |
10 mg | |
O=C(O)C[C@]1(O)C(O[C@H](CCCCCCC2=CC=C(Cl)C=C2Cl)C1)=O | |
389.27 | |
Research | |
Metabolism-sugar/lipid metabolism |