missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Selenite, purum p.a, ≥96.0% (RT), Honeywell™
Purum p.a.,≥96.0% (RT)
62.79€ - 261.59€
Identificatori chimici
CAS | 7782-82-3 |
---|---|
Formula molecolare | Na2O3Se |
Molecular Weight (g/mol) | 172.95 |
Numero MDL | MFCD00003525 |
InChI Key | BVTBRVFYZUCAKH-UHFFFAOYSA-L |
Sinonimo | sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide |
PubChem CID | 23669629 |
IUPAC Name | sodium;hydrogen selenite |
SMILES | [Na+].[Na+].[O-][Se]([O-])=O |
Descrizione
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Identificatori chimici
7782-82-3 | |
172.95 | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
23669629 | |
[Na+].[Na+].[O-][Se]([O-])=O |
Na2O3Se | |
MFCD00003525 | |
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide | |
sodium;hydrogen selenite |
Specifica
Sodium hydrogen selenite | |
Na2O3Se | |
MFCD00003525 | |
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide | |
[Na+].[Na+].[O-][Se]([O-])=O | |
172.95 | |
150.96g/mol |
7782-82-3 | |
NaHSeO3 | |
UN2630 | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
sodium;hydrogen selenite | |
23669629 | |
≥96.0% (RT) |
Sicurezza e movimentazione
P261-P273-P301 + P310-P311-P501
missing translation for 'dotInformation' : 6.1 / PGI
missing translation for 'rtecsNumber' : 231-966-3