Learn More
Metro™ Stainless Lab Worktable, Stainless Island Top and 3-Sided Frame
Keep lab materials clean and safe. Metro Stainless Lab Worktable features Type 304 stainless steel construction throughout and 3-sided Frame providing corrosion-proof performance and stability you can count on.
Marca:  Metro™ LTSM36UIS
Dettagli aggiuntivi : Peso : 25.40000kg
Codice prodotto. 15957131
  • 1900.00€ / Pezzo
Spedizione stimata: 20-12-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifica
Specifica
35.75 in.
35.75 x 30 x 35 in. (90.8 x 76.2 x 89 cm)
89 cm
3 Sided frame, adaptor insert
273 kg
90.8 cm
Stainless Steel
Mobile Table
Ideal for lab
35 in.
30 in.
76.2 cm
600 lb.