Learn More
Buerkle™ SteriBag™ StandUp Polyethylene Sampling Bags
Buerkle™ SteriBag™ StandUp Polyethylene Sampling Bags are sterile sample bag with a stand-up base.
Marca: Buerkle™ 5344-0600
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Stands stably
- Protected against discharge
- Sterilised
- With labelling field
Specifica
Sample Bag | |
Clear | |
Polyethylene | |
500 | |
500/Pk. | |
229 mm | |
229 mm |
600 mL | |
Ideal for Samples Available as Liquids, Powders, Solids and Pastes, Suitable for Foods and Can be Frozen | |
Sterile | |
Tight closing | |
229 x 152 mm | |
152 mm |