Learn More
STIP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA589626
338.68 EUR valido fino al 2025-03-21
Usa il codice promo "24111" per avere il tuo prezzo promozionale.
Allerta:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Descrizione
Immunogen sequence: MEQVNELKEK GNKALSVGNI DDALQCYSEA IKLDPHNHVL YSNRSAAYAK KGDYQKAYED GCKTVDLKPD WGKGYSRKAA ALEFLNRFEE AKRTYEEGLK HEANNPQLKE GLQNMEARLA ERKFMNPFNM PNLYQKLESD PRTRTLLSDP TYRELIEQLR NKPSDLGTKL QDPRIMTTLS VLLGVDLGSM DEEEEIATPP PPPPPKKETK PEPMEEDLPE NKKQALKEKE LGNDAYKKKD FDTALKHYDK AKELDPTNMT YITNQAAVYF EKGDYNKCRE LCEKAIEVGR ENREDYRQIA; Positive Samples: HeLa, COS-7, COS-1, OVCAR3, Mouse brain; Cellular Location: Cytoplasm, Nucleus
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
Specifica
STIP1 | |
Polyclonal | |
Unconjugated | |
STIP1 | |
epididymis secretory sperm binding protein Li 94n, HEL-S-94n, HOP, hsc70/Hsp90-organizing protein, Hsp70/Hsp90 organizing protein, Hsp70/Hsp90-organizing protein, IEF SSP 3521, IEF-SSP-3521, mSTI1, NY-REN-11 antigen, P60, Renal carcinoma antigen NY-REN-11, STI1, STI1L, Stip1, stress induced phosphoprotein 1, stress-induced phosphoprotein 1, stress-induced-phosphoprotein 1, stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein), stress-inducible protein, transformation-sensitive protein IEF SSP 3521 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10963, 192277, 20867 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
O35814, P31948, Q60864 | |
STIP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human STIP1 (NP_0068101) | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.