Learn More
STK25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA597240
336.01 EUR valido fino al 2025-03-29
Usa il codice promo "24111" per avere il tuo prezzo promozionale.
Allerta:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Descrizione
Immunogen sequence: EESSSEDSDI DGEAEDGEQG PIWTFPPTIR PSPHSKLHKG TALHSSQKPA EPVKRQPRSQ CLSTLVRPVF GELKEKHKQS GGSVGALEEL ENAFSLAEES CPGISDKLMV HLVERVQRFS HNRNHLTSTR; Positive Samples: SW620, Mouse small intestine; Cellular Location: Cytoplasm, Golgi apparatus
STK25 (YSK1) is a serine/threonine kinase that belongs to the GCK family of kinases. It has been implicated as an upstream regulator of MAP kinase signaling pathways. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifica
STK25 | |
Polyclonal | |
Unconjugated | |
STK25 | |
1500019J11Rik, AU018434, C86992, hypothetical protein, serine/threonine kinase 25, serine/threonine kinase 25 (STE20 homolog, yeast), serine/threonine kinase 25 (Ste20, yeast homolog), serine/threonine kinase 25 (yeast), serine/threonine-protein kinase 25, SOK1, SOK-1, Ste20, yeast homolog, ste20/oxidant stress response kinase 1, Ste20-like, ste20-like kinase, sterile 20/oxidant stress-response kinase 1, STK25, STK25 protein, Yeast Sps1/Ste20-related kinase 1, Ysk1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10494, 59041 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
O00506, Q9Z2W1 | |
STK25 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 297-426 of human STK25 (NP_0063652) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.