Learn More
Bohlender™ BOLA™ PTFE Y-shaped Tubing Connectors
Connect elastic tubing with these tubing connectors. BOLA PTFE Tubing Connectors feature universal chemical resistance and are vacuum suitable.
Marca: Bohlender™ D576-06
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- PTFE Y-shaped fitting
- Three connectors for elastic tubing (e.g. Viton, Tygon, silicone)
- Universal chemical resistance
- Product is only exposed to PTFE
Compliance
FDA
Specifica
FDA | |
Autoclavable | |
PTFE | |
53 mm | |
-200°C | |
5 x 9mm (I.D. x O.D.) | |
Y | |
250°C |
Tubing Connector | |
5 mm | |
9 mm | |
Elastic tubing | |
10/Pk. | |
UNF 1/4 in.; 28 G | |
White |