Learn More
Lenz Laborglasinstrumente™ Teste di distillazione dritte
For placement onto distillation flasks for separation of drops. Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight, are manufactured from durable Duran™ glass tubing.
Marca: Lenz Laborglasinstrumente™ 05035011
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Straight with NS cone and socket
- Temperature and chemical resistance
- Minimal thermal expansion
Specifica
For separation of drops (Direct placement onto distillation flasks) | |
Straight Splash Heads | |
Duran Glass | |
NS 14/23 NS 14/23 |