Learn More
Sucrose Brix/RI Standards, Paragon Scientific™
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
Marca:  Paragon Scientific SS125
Dettagli aggiuntivi : Peso : 0.20000kg
Codice prodotto. 16040410
  • 56.30€ / 15mL
Spedizione stimata: 04-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Specifica
Specifica
12.50 Brix
15 mL
Offerte speciali
Nominal values quoted