Learn More
3M™ Swab-Sampler
Convenient and quick collection device for environmental sampling and testing over small surface areas.
Marca:  3M™ 7000030300
Dettagli aggiuntivi : Peso : 0.48500kg
Codice prodotto. 17238520
  • 151.58€ / pezzi 100
Spedizione stimata: 10-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Letheen Broth vial is ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifica
Specifica
4 mL Letheen Broth
100 /Cs.
Swab