Learn More
Tocris Bioscience™ TH 9619 disodium salt
MTHFD2 inhibitor
Marca: Tocris Bioscience™ 7719/10
Dettagli aggiuntivi : Peso : 0.00970kg
Descrizione
TH 9619 disodium salt is a potent and selective methylenetetrahydrofolate dehydrogenase inhibitor (MTHFD2; IC50 = 47 nM). It shows efficacity on HL-60 cell viability with EC50 of 11nM. In vitro, It shows antiproliferative efficacy in acute myeloid leukemia (AmL) cells and T-ALL Jurkat cells. In vivo, TH9619 disodium salt impairs cancer progression in a mouse disease model of AmL.Specifica
Disodium N-[[5-[[2-(2,4-Diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)acetyl]amino]-3-fluoro-2-pyridinyl]carbonyl]-L-glutamate | |
Dehydrogenase Inhibitors | |
0.98 |
10 mg | |
C17H16FN7Na2O7 |