Learn More
Abeba™ The Original 7080 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7080/36
Additional Details : Peso : 0.50000kg
Description
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather, perforated
- Light blue/White
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
Light Blue, White | |
36 | |
7080 |