Learn More
3M™ MonitorMark™ Time Temperature Indicator
Solves the challenge of monitoring thermal exposure of temperature-sensitive products during transportation and storage.
Marca:  3M™ MonitorMark™ 7000001929
Dettagli aggiuntivi : Peso : 0.48500kg
Codice prodotto. 17278510
  • 3726.17€ / pezzi 500
Spedizione stimata: 19-09-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Inexpensive solution for monitoring product exposure to temperatures above the specified limit over a 48 hour time period.
  • Self-adhesive backing for easy attachment to secondary packaging.
  • Easily-interpreted visual results.
  • Results indicate both exposure and relative time over which exposure occurred.
  • Removable film activation strip starts the monitoring process on demand for accuracy.
Specifica
Specifica
500 /Cs.
48 Hours
72°F
22°C