Learn More
SI Analytics™ Titration Tip
SI Analytics™ Titration Tip is designed to be attached to a titrator to increase precision in titration application.
Marca: SI Analytics™ 285227263
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Tip for titrator
- For use with TITRONIC™ Series and TitroLine™ Series
Specifica
Blue | |
0.7 mm | |
Titration Tip | |
TITRONIC™ Basic, TitroLine™ Easy and TA 05 To TA 50 plus |