Learn More
Ansell™ Guanti in nitrile verde TouchNTuff™ serie 93-300
Conformi con ASTM D 3577, tipo II. I guanti Ansell™ TouchNTuff™ in nitrile verde serie 93-300 sono ideali per l'uso durante la produzione di componenti elettronici e semiconduttori e nei test di laboratorio.
Marca: Ansell™ 93-300_8.5-9
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove
- Provides the wearer with a high level of comfort and increased protection in the forearm (longer cuff)
- Manufactured from nitrile, offering three times the puncture resistance of standard latex gloves
- Made without natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type l) wearers
- Suitable for ISO Class 5 or higher clean room environments
Specifica
Nitrile | |
Nitrile | |
Yes | |
8.5 | |
0.12 mm | |
For blending of liquids, filtration processes, vaccines injectables operations etc. | |
Splash and high puncture resistance |
Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II | |
Nitrile | |
Gloves | |
93-300 | |
Textured fingertips | |
240 Pairs | |
93-300 |