Learn More
Köttermann Underbench Acid and Base Cabinet
Constructed with durable polypropylene, this Underbench Acid and Base Cabinet is built to endure the aggressive conditions produced by corrosive acid fumes. Liquid-tight pull-out trays offer easy access while minimizing health and safety risks.
Marca: Köttermann 377-0008
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
- Cabinet carcasses and pull-out trays made of resistant polypropylene
- Liquid-tight drawers on low-friction PTFE strips with pull-out stop
- Removal of aggressive vapors via optional ventilation with an external fan
- Corrosion-resistant locks and hinges
- Ideal for installation under high-performance fume cupboards
Specifica
2 Doors, 4 Drawers | |
Hinged, Drawer | |
No Ventilation Fan | |
Polypropylene |
515 x 1100 x 720 mm | |
Acid and Base Storage | |
White |