Learn More
Abeba™ UNI6 1781 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Marca: Abeba™ 1781/39
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- A-micro
- Black
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver Point
- Instep area with lacing
- **Size 35 with replaceable acc Wave insole (art. 3576)
Specifica
CE EN ISO 20345:2011 S2 SRC | |
A-micro | |
Slip-resistant | |
1781 |
Black | |
39 | |
Polyurethane | |
Steel |