Learn More
ABEBA™ Uni6 Black Safety Trainers S3 ESD
Breathable, water repellent trainers made of knitted fabric, slip resistant PU/TPU outsole, steel toe cap, textile antipuncture
Marca: ABEBA™ 31676/40
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Made for safety and comfort
- Comply to EN ISO 20345: 2012, S3, SRC
- ESD dissipative according EN 61340
- Slip resistant PU/TPU outsole
- Breathable and water repellent knitted fabric
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Breathable and antibacterial lining with silver ions
- Removable insole active comfort
Specifica
Black | |
40 | |
ESD according 61340 class climatic 1, EN ISO 20345: 2012 S3 SRC | |
Trainers | |
S3 SRC ESD |
Knitted Fabric | |
Steel | |
Unisex | |
Water Repellent |