Learn More
Kinesis™ PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Marca: Kinesis™ 02006-30
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
- Biocompatible, inert and easy to cut
- Great for high-pressure applications
- Wide choice of outer and inner diameters
- Many sizes color-coded for easier inner diameter identification
Specifica
Tubing | |
High-Pressure Applications | |
5 ft. | |
7000 psi (483 bar) |
Green | |
0.03 in. | |
PEEK |