missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Semi-Rigid Tubing, UNF(M)
VapLock™ Adapters for Semi-Rigid Tubing, UNF(M) are designed for secure liquid-tight connections
Marca: Cole-Parmer™ 12020-03
53.68 EUR valido fino al 2024-09-17
Usa il codice promo "21690" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Color-coded for easy identification
- Features two-piece nut and ferrule system
Servizi
None
Specifica
Solvent Safety | |
Tubing Adapter, 1/16 in. OD x 1/4-28 UNF(M), green PP w/ blue ETFE; 10/Pk. | |
VapLock Solvent Containment Systems |
1/16 in. OD Compression x UNF(M) | |
Green / Blue PP / ETFE | |
1/4 in.-28 UNF(M) |