Learn More
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings
For the disintegration of COD, total nitrogen and total phosphorus
Marca: WTW™ 1P23-1
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Standardized digestion programs for the standard parameters COD, quick COD (148°C, 20 min.), total nitrogen, total phosphorus, TOC and heavy metals of galvanizing with automatic heating and switch-off. 8 user-defined digestions at temperatures between 25-170°C can be programmed for special degradations and therefore also allow high temperature digestions. Two different programs for 12 cuvettes each can be run at the same time. You can use an external temperature sensor instead of a vial with data output to the PC/printer for analytical quality assurance (AQA).Specifica
Thermoreactor | |
230/115 V | |
315 mm | |
Routine and specialized tasks in wastewater and in laboratories | |
2 x 12 Reaction Cuvettes in 2 heating blocks |
50/60 Hz | |
560 W | |
115/230 V ±15%, 50/60 Hz, 560 W | |
185 mm | |
256 mm |