Learn More
VACUUBRAND™ VARIO™ PC 3010 NT Select Chemistry Pumping Unit
The PC 3010 NT pumping unit precisely controls vacuum level to achieve unparalleled process control. With its combination of high pumping speed and fractional-millibar ultimate vacuum, this pump is suitable for demanding applications, even with high boiling point solvents at low temperatures.
Marca: VACUUBRAND™ 25744851
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in laboratory
- Automatic boiling point detection and adjustment of vacuum for short process times
- Ideal for evaporation of large amounts of high-boiling solvents, even at low temperatures
- Extra long diaphragm life for minimum operational and servicing costs
- Environmental compatibility through low energy consumption and efficient solvent recovery
Specifica
0.6 mbar | |
47 dBA | |
29.7 kg | |
450 mm | |
387 mm | |
IP40 | |
1.1 bar | |
200/230 V |
Mounted Pump, Manual | |
CH | |
12.8 m3/hr. | |
616 mm | |
Vacuum System | |
Work in Miniplants, Kilo Labs, Scale-up Facilities | |
Rotary Evaporators, Vacuum Drying Ovens, Vacuum Distillation Columns |